Synthware Replacement T-Bore Glass Stopcocks | Avantor
Synthware Replacement T-Bore Glass Stopcocks
Replacement T-bore glass stopcock, 4 mm
Catalog # 60005-182
Supplier:   Kemtech America
Fournisseur #:  S511740
Synthware Replacement T-Bore Glass Stopcocks
Catalog # 60005-182
Supplier:  Kemtech America
Synthware Replacement T-Bore Glass Stopcocks
Synthware Replacement T-Bore Glass Stopcocks
Replacement T-bore glass stopcock, 4 mm
Price
44,41 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Glass plug ONLY.
  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands
For use with Kemtech S69 series stainless steel retention clips.
Spécifications
  • Désignation:
    Replacement T-bore glass stopcock
  • Bore Size:
    4 mm