Synthware Replacement T-Bore Glass Stopcocks
Replacement T-bore glass stopcock, 2 mm
Catalog # 60005-180
Supplier: Kemtech America
Fournisseur #: S510217
Jump to:
Product Details & Documents
Glass plug ONLY.
- Heavy wall construction
- 33 expansion borosilicate glass
- Standard ground glass joints work with other brands
For use with Kemtech S69 series stainless steel retention clips.
Spécifications
- Désignation:Replacement T-bore glass stopcock
- Bore Size:2 mm