[ST] Joint Clips
Plastic joint clip, 34/45
Catalog # 60004-954
Supplier: Kemtech America
Fournisseur #: CS003445
Jump to:
Product Details & Documents
Made of plastic.
- Heavy duty plastic construction
- Color coded
- Fit standard taper joints
Our heavy duty keck clips are perfect for all standard taper ground glass joints.
Spécifications
- Désignation:Plastic joint clip
- ST Joint Size:34/45