Synthware Springs
6.35 mm, Spring, Stainless steel
Catalog # 60004-940
Supplier: Kemtech America
Supplier #: TH006
Jump to:
Product Details & Documents
Stainless steel retention springs.
- Chemically resistant stainless steel
- Compatible with other brands
Stainless steel springs for use with glassware that features hooks.
Specifications
- Désignation:Spring
- Longueur:6.35 mm
- Matière:Stainless steel