Synthware Springs
Synthware Springs
6.35 mm, Spring, Stainless steel
Catalog # 60004-940
Supplier:   Kemtech America
Supplier #:  TH006
Synthware Springs
Catalog # 60004-940
Supplier:  Kemtech America
Synthware Springs
Synthware Springs
6.35 mm, Spring, Stainless steel
Price
$3.52
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Stainless steel retention springs.
  • Chemically resistant stainless steel
  • Compatible with other brands
Stainless steel springs for use with glassware that features hooks.
Specifications
  • Désignation:
    Spring
  • Longueur:
    6.35 mm
  • Matière:
    Stainless steel