Synthware PTFE Stoppers, Solid and Hollow
24/40
Catalog # 60004-906
Supplier: Kemtech America
Supplier #: S532440
Jump to:
Product Details & Documents
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
- Extraction nut
- Teflon® construction
Available in solid or hollow formats.
Specifications
- ST Joint No.:24/40