Synthware PTFE Stoppers, Solid and Hollow
Synthware PTFE Stoppers, Solid and Hollow
19/22
Catalog # 60004-904
Supplier:   Kemtech America
Supplier #:  S531922
Synthware PTFE Stoppers, Solid and Hollow
Catalog # 60004-904
Supplier:  Kemtech America
Synthware PTFE Stoppers, Solid and Hollow
Synthware PTFE Stoppers, Solid and Hollow
19/22
Price
$52.53
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
  • Extraction nut
  • Teflon® construction
Available in solid or hollow formats.
Specifications
  • ST Joint No.:
    19/22