Synthware PTFE Stoppers, Solid and Hollow
19/22
Catalog # 60004-904
Supplier: Kemtech America
Supplier #: S531922
Jump to:
Product Details & Documents
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
- Extraction nut
- Teflon® construction
Available in solid or hollow formats.
Specifications
- ST Joint No.:19/22