Synthware PTFE Stoppers, Solid and Hollow | Avantor
Synthware PTFE Stoppers, Solid and Hollow
10/18
Catalog # 60004-900
Supplier:   Kemtech America
Fournisseur #:  S531018
Synthware PTFE Stoppers, Solid and Hollow
Catalog # 60004-900
Supplier:  Kemtech America
Synthware PTFE Stoppers, Solid and Hollow
Synthware PTFE Stoppers, Solid and Hollow
10/18
Price
$47.67
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
  • Extraction nut
  • Teflon® construction
Available in solid or hollow formats.
Spécifications
  • ST Joint No.:
    10/18