Synthware PTFE Stoppers, Solid and Hollow
10/18
Catalog # 60004-900
Supplier: Kemtech America
Fournisseur #: S531018
Jump to:
Product Details & Documents
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
- Extraction nut
- Teflon® construction
Available in solid or hollow formats.
Spécifications
- ST Joint No.:10/18