Synthware PTFE Stoppers
Synthware PTFE Stoppers
29/42
Catalog # 60004-868
Supplier:   Kemtech America
Supplier #:  S112942
Synthware PTFE Stoppers
Catalog # 60004-868
Supplier:  Kemtech America
Synthware PTFE Stoppers
Synthware PTFE Stoppers
29/42
Price
$153.67
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Stoppers feature PTFE handles for easy gripping.
  • T-handle design
  • Teflon® construction
Teflon® handle stoppers for easy griping.
Specifications
  • ST Joint No.:
    29/42