Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Synthware Storage Vessel, #15 O-Ring Joint

Synthware Storage Vessel, #15 O-Ring Joint

50 ml, Storage vessel, #15 O-ring joint, 190 mm high, 0 - 4 mm
Catalog # 60004-686
Supplier:   Kemtech America
Supplier #:  S490050
Synthware Storage Vessel, #15 O-Ring Joint
Catalog # 60004-686
Supplier:  Kemtech America
Synthware Storage Vessel, #15 O-Ring Joint
Synthware Storage Vessel, #15 O-Ring Joint

50 ml, Storage vessel, #15 O-ring joint, 190 mm high, 0 - 4 mm

Price
$238.56
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Storage vessel with a #15 O-ring joint on the sidearm and a high vacuum valve to control gas or liquid flow into the vessel.

  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands

Supplied complete with one #116 Viton® O-ring, and one high vacuum valve.

Specifications
  • Capacité:
    50 ml
  • Désignation:
    Storage vessel, #15 O-ring joint, 190 mm high
  • Valve Size:
    0 - 4 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.