Synthware Flask, Filtering, Microscale, Kemtech America
Synthware Flask, Filtering, Microscale, Kemtech America
25 ml
Catalog # 60004-224
Supplier:   Kemtech America
Supplier #:  F150025
Synthware Flask, Filtering, Microscale, Kemtech America
Catalog # 60004-224
Supplier:  Kemtech America
Synthware Flask, Filtering, Microscale, Kemtech America
Synthware Flask, Filtering, Microscale, Kemtech America
25 ml
Price
47,05 $
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Filtering flask for microscale applications.
  • Microscale
  • Heavy wall Borosilicate construction
  • Beaded top rim
  • Barbed side arm
Top is tooled for a #2 neoprene filter adapter or #3 rubber stopper.
Specifications
  • Capacité:
    25 ml