Synthware Flask, Filtering, Microscale, Kemtech America
25 ml
Catalog # 60004-224
Supplier: Kemtech America
Supplier #: F150025
Synthware Flask, Filtering, Microscale, Kemtech America
Catalog # 60004-224
Supplier: Kemtech America
Jump to:
Product Details & Documents
Filtering flask for microscale applications.
- Microscale
- Heavy wall Borosilicate construction
- Beaded top rim
- Barbed side arm
Top is tooled for a #2 neoprene filter adapter or #3 rubber stopper.
Specifications
- Capacité:25 ml