Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
10 ml, 14/10
Catalog # 60004-220
Supplier:   Kemtech America
Supplier #:  F501010
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
Catalog # 60004-220
Supplier:  Kemtech America
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
10 ml, 14/10
Price
$44.61
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Minimum order quantity is 1
Product Details & Documents
Erlenmeyer threaded flask for microscale applications.
  • Heavy wall borosilicate construction
  • Threaded upper joint
  • Open cap included
Supplied with a nylon compression cap, Viton® O-ring, and PTFE-lined silicone septum.
Spécifications
  • ST Joint No.:
    14/10
  • Capacité:
    10 ml
Accessories
Informations sur le produit
  • Catalog #
    I.D.×W
    O-Ring Size
    Availability
    Price
  • Viton® O-Ring, 14/10 threaded joints
    Viton® O-Ring, 14/10 threaded joints
    60004-998
    014
    Log in pour la disponibilité
    Each - $5.61
Informations sur le produit
  • Catalog #
    Material
    Cap Size
    Availability
    Price
  • Nylon cap
    Nylon cap
    60005-054
    Nylon
    20 mm
    Log in pour la disponibilité
    Each - $4.65