Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
25 ml, 14/10
Catalog # 60004-186
Supplier:   Kemtech America
Supplier #:  F301025
Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
Catalog # 60004-186
Supplier:  Kemtech America
Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
25 ml, 14/10
Price
$60.68
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Supplied with one compression cap, one Viton® O-ring, and one fluoropolymer resin-faced silicone rubber septum.
  • Threaded neck
  • Ground glass joint
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
  • Cap included
Threaded ground glass joint allows for the use of a threaded cap or a ground glass stopper.
Ordering information: For replacement caps, contact your Avantor representative.
Specifications
  • ST Joint No.:
    14/10
  • Capacité:
    25 ml