Synthware Nylon Caps
Nylon cap, 14 mm
Catalog # 60003-754
Supplier: Kemtech America
Fournisseur #: GL14
Jump to:
Product Details & Documents
Replacement caps are designed for use with solid phase reaction vessels.
Nylon construction
- Closed top
Spécifications
- Taille du bouchon:14 mm
- Désignation:Nylon cap