Synthware Round-Bottom Flasks with Four Necks
2000 ml, 29/42, 24/40
Catalog # 60002-998
Supplier: Kemtech America
Fournisseur #: F21842L
Jump to:
Product Details & Documents
With identical vertical side joints.
- 3 Straight side necks
- Ground glass joints
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
Spécifications
- Capacité:2000 ml
- Center Neck [ST] Joint:29/42
- Side Neck ST Joints:24/40