Synthware Round-Bottom Flasks with Four Necks
1000 ml, 24/40, 24/40
  60002-982
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  F214000
Synthware Round-Bottom Flasks with Four Necks
  60002-982
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Synthware Round-Bottom Flasks with Four Necks
Synthware Round-Bottom Flasks with Four Necks
1000 ml, 24/40, 24/40
$405.07
Each
With identical vertical side joints.
  • 3 Straight side necks
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
  • Capacité:
    1000 ml
  • Center Neck [ST] Joint:
    24/40
  • Side Neck ST Joints:
    24/40
  • Cat. No.:
    60002-982