Synthware Round-Bottom Flasks with Four Necks
1000 ml, 24/40, 24/40
60002-982
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
: F214000
Synthware Round-Bottom Flasks with Four Necks
60002-982
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
With identical vertical side joints.
- 3 Straight side necks
- Ground glass joints
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
- Capacité:1000 ml
- Center Neck [ST] Joint:24/40
- Side Neck ST Joints:24/40
- Cat. No.:60002-982