Synthware Round-Bottom Flasks with Four Necks
Synthware Round-Bottom Flasks with Four Necks
500 ml, 29/42, 29/42
Catalog # 60002-974
Supplier:   Kemtech America
Supplier #:  F218500
Synthware Round-Bottom Flasks with Four Necks
Catalog # 60002-974
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Four Necks
Synthware Round-Bottom Flasks with Four Necks
500 ml, 29/42, 29/42
Price
$333.49
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
With identical vertical side joints.
  • 3 Straight side necks
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
Specifications
  • Capacité:
    500 ml
  • Center Neck [ST] Joint:
    29/42
  • Side Neck ST Joints:
    29/42