Synthware Round-Bottom Flasks with Four Necks
500 ml, 29/42, 29/42
Catalog # 60002-974
Supplier: Kemtech America
Supplier #: F218500
Jump to:
Product Details & Documents
With identical vertical side joints.
- 3 Straight side necks
- Ground glass joints
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
Specifications
- Capacité:500 ml
- Center Neck [ST] Joint:29/42
- Side Neck ST Joints:29/42