Synthware Round-Bottom Flasks with Four Necks
250 ml, 24/40, 24/40
60002-962
: F214250
With identical vertical side joints.
- 3 Straight side necks
- Ground glass joints
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
- Capacité:250 ml
- Center Neck [ST] Joint:24/40
- Side Neck ST Joints:24/40