Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
2000 ml, 45/50, 24/40
  60002-778
Supplier #:  F43542L
Synthware Round-Bottom Flasks with Three Necks, Angled
  60002-778
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
2000 ml, 45/50, 24/40
$320.54
Each
Minimum order quantity is 1
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
  • Capacité:
    2000 ml
  • Center Neck [ST] Joint:
    45/50
  • Side Neck ST Joints:
    24/40