Synthware Round-Bottom Flasks with Three Necks, Angled
250 ml, 24/40, 24/40
  60002-726
 :  F434250
Synthware Round-Bottom Flasks with Three Necks, Angled
  60002-726
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
250 ml, 24/40, 24/40
$220.20
Each
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
  • Capacité:
    250 ml
  • Center Neck [ST] Joint:
    24/40
  • Side Neck ST Joints:
    24/40