Synthware Round-Bottom Flasks with Three Necks, Angled
100 ml, 24/40, 24/40
Catalog # 60002-716
Supplier: Kemtech America
Fournisseur #: F434400
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 60002-716
Supplier: Kemtech America
Jump to:
Product Details & Documents
Round bottom with [ST] joints.
- 3 Angled necks
- Ground glass joints
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Spécifications
- Capacité:100 ml
- Center Neck [ST] Joint:24/40
- Side Neck ST Joints:24/40