Synthware Round-Bottom Flasks with Three Necks, Angled | Avantor
Synthware Round-Bottom Flasks with Three Necks, Angled
100 ml, 14/20, 14/20
Catalog # 60002-706
Supplier:   Kemtech America
Fournisseur #:  F431100
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 60002-706
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
100 ml, 14/20, 14/20
Price
$174.19
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Spécifications
  • Capacité:
    100 ml
  • Center Neck [ST] Joint:
    14/20
  • Side Neck ST Joints:
    14/20