Synthware Round-Bottom Flasks with Three Necks, Angled
50 ml, 24/40, 24/40
Catalog # 60002-704
Supplier:   Kemtech America
 :  F434450
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 60002-704
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
50 ml, 24/40, 24/40
Price
$195.30
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Spécifications
  • Capacité:
    50 ml
  • Center Neck [ST] Joint:
    24/40
  • Side Neck ST Joints:
    24/40