Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
50 ml, 24/40, 14/20
  60002-700
Supplier #:  F434150
Synthware Round-Bottom Flasks with Three Necks, Angled
  60002-700
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
50 ml, 24/40, 14/20
$141.17
Each
Minimum order quantity is 1
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
  • Capacité:
    50 ml
  • Center Neck [ST] Joint:
    24/40
  • Side Neck ST Joints:
    14/20