Synthware Round-Bottom Flasks with Three Necks, Angled
50 ml, 14/20, 14/20
Catalog # 60002-694
Supplier: Kemtech America
Fournisseur #: F431150
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 60002-694
Supplier: Kemtech America
Jump to:
Product Details & Documents
Round bottom with [ST] joints.
- 3 Angled necks
- Ground glass joints
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Spécifications
- Capacité:50 ml
- Center Neck [ST] Joint:14/20
- Side Neck ST Joints:14/20
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.