Synthware Round-Bottom Flasks with Three Necks, Angled
25 ml, 14/20, 14/20
  60002-688
 :  F431125
Synthware Round-Bottom Flasks with Three Necks, Angled
  60002-688
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
25 ml, 14/20, 14/20
134,14 $
Each
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
  • Capacité:
    25 ml
  • Center Neck [ST] Joint:
    14/20
  • Side Neck ST Joints:
    14/20