Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
1000 ml, 24/40, 24/40
60002-658
: F41241L
Round-bottom flasks with one straight [ST] joint and one angled [ST] joint
- Angled side neck
- Heavy wall borosilicate construction
- Ground glass joint
- Various sizes available
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
- Capacité:1000 ml
- Center Neck [ST] Joint:24/40
- Side Neck ST Joints:24/40