Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
1000 ml, 24/40, 24/40
  60002-658
 :  F41241L
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
  60002-658
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
1000 ml, 24/40, 24/40
207,40 $
Each
Round-bottom flasks with one straight [ST] joint and one angled [ST] joint
  • Angled side neck
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Various sizes available
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
  • Capacité:
    1000 ml
  • Center Neck [ST] Joint:
    24/40
  • Side Neck ST Joints:
    24/40