Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
250 ml, 19/22, 19/22
Catalog # 60002-622
Supplier: Kemtech America
Supplier #: F419250
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Catalog # 60002-622
Supplier: Kemtech America
Jump to:
Product Details & Documents
Round-bottom flasks with one straight [ST] joint and one angled [ST] joint
- Angled side neck
- Heavy wall borosilicate construction
- Ground glass joint
- Various sizes available
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Spécifications
- Capacité:250 ml
- Center Neck [ST] Joint:19/22
- Side Neck ST Joints:19/22