Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
250 ml, 19/22, 19/22
Catalog # 60002-622
Supplier:   Kemtech America
Supplier #:  F419250
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Catalog # 60002-622
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
250 ml, 19/22, 19/22
Price
$110.04
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Minimum order quantity is 1
Product Details & Documents
Round-bottom flasks with one straight [ST] joint and one angled [ST] joint
  • Angled side neck
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Various sizes available
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Spécifications
  • Capacité:
    250 ml
  • Center Neck [ST] Joint:
    19/22
  • Side Neck ST Joints:
    19/22