Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
100 ml, 14/20, 14/20
Catalog # 60002-606
Supplier: Kemtech America
Fournisseur #: F411100
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Catalog # 60002-606
Supplier: Kemtech America
Jump to:
Product Details & Documents
Round-bottom flasks with one straight [ST] joint and one angled [ST] joint
- Angled side neck
- Heavy wall borosilicate construction
- Ground glass joint
- Various sizes available
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Spécifications
- Capacité:100 ml
- Center Neck [ST] Joint:14/20
- Side Neck ST Joints:14/20