Synthware Flat-Bottom Flasks, Kemtech America
1 L, 29/42
60002-248
: F128000
Single-neck, flat-bottom flasks with an outer joint.
- Single neck
- Ground glass joint
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
- Flat bottom
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
- ST Joint No.:29/42
- Capacité:1 L