Synthware Flat-Bottom Flasks, Kemtech America
300 ml, 24/40
60002-238
: F124300
Single-neck, flat-bottom flasks with an outer joint.
- Single neck
- Ground glass joint
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
- Flat bottom
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
- ST Joint No.:24/40
- Capacité:300 ml