Synthware Flat-Bottom Flasks, Kemtech America
300 ml, 24/40
  60002-238
 :  F124300
Synthware Flat-Bottom Flasks, Kemtech America
  60002-238
Synthware Flat-Bottom Flasks, Kemtech America
Synthware Flat-Bottom Flasks, Kemtech America
300 ml, 24/40
$72.49
Each
Single-neck, flat-bottom flasks with an outer joint.
  • Single neck
  • Ground glass joint
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
  • Flat bottom
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
  • ST Joint No.:
    24/40
  • Capacité:
    300 ml