Synthware Extraction Thimbles, Glass
Medium extractor size, 35×90 mm, Extra coarse
60002-220
: E113590X
All-glass extraction thimbles permit viewing extraction process and subsequent weighing.
- Heavy wall borosilicate construction
- Coarse or extra coarse porosity available
: Small thimble is used with condenser 60002-198 and extractor 60002-204; Medium thimble is used with condenser 60002-200 and extractors 60002-206 or 60002-208; Large thimble is used with condenser 60002-202 and extractors 60002-210 or 60002-212.
- Porosité:Extra coarse
- O.D.×H:35×90 mm
- Fits:Medium extractor size