Synthware Extraction Thimbles, Glass
Synthware Extraction Thimbles, Glass
Small extractor size, 25×85 mm, Coarse
Catalog # 60002-214
Supplier:   Kemtech America
Supplier #:  E112585C
Synthware Extraction Thimbles, Glass
Catalog # 60002-214
Supplier:  Kemtech America
Synthware Extraction Thimbles, Glass
Synthware Extraction Thimbles, Glass
Small extractor size, 25×85 mm, Coarse
Price
$107.45
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
All-glass extraction thimbles permit viewing extraction process and subsequent weighing.
  • Heavy wall borosilicate construction
  • Coarse or extra coarse porosity available
Ordering information: Small thimble is used with condenser 60002-198 and extractor 60002-204; Medium thimble is used with condenser 60002-200 and extractors 60002-206 or 60002-208; Large thimble is used with condenser 60002-202 and extractors 60002-210 or 60002-212.
Specifications
  • Porosité:
    Coarse
  • O.D.×H:
    25×85 mm
  • Fits:
    Small extractor size