Synthware Distillation Connecting Adapters
24/40, [ST] 14/20, 19/22
Catalog # 60000-606
Supplier:   Kemtech America
 :  A354914
Synthware Distillation Connecting Adapters
Catalog # 60000-606
Supplier:  Kemtech America
Synthware Distillation Connecting Adapters
Synthware Distillation Connecting Adapters
24/40, [ST] 14/20, 19/22
Price
$102.53
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
  • Inner joints at bottom and side arm
  • Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
Specifications
  • Bottom [ST] Joint Size:
    24/40
  • Top ST Joint Size:
    [ST] 14/20
  • Side ST Joint Size:
    19/22