Synthware Distillation Connecting Adapters | Avantor
Synthware Distillation Connecting Adapters
24/40, [ST] 14/20, 24/40
Catalog # 60000-604
Supplier:   Kemtech America
Fournisseur #:  A351424
Synthware Distillation Connecting Adapters
Catalog # 60000-604
Supplier:  Kemtech America
Synthware Distillation Connecting Adapters
Synthware Distillation Connecting Adapters
24/40, [ST] 14/20, 24/40
Price
101,71 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
  • Inner joints at bottom and side arm
  • Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
Spécifications
  • Bottom [ST] Joint Size:
    24/40
  • Top ST Joint Size:
    [ST] 14/20
  • Side ST Joint Size:
    24/40