Synthware Distillation Connecting Adapters
24/40, [ST] 24/40, 24/40
Catalog # 60000-600
Supplier: Kemtech America
Supplier #: A352440
Jump to:
Product Details & Documents
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
- Inner joints at bottom and side arm
- Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
Specifications
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 24/40
- Side ST Joint Size:24/40