Synthware Distillation Connecting Adapters
19/22, [ST] 19/22, 19/22
Catalog # 60000-598
Supplier: Kemtech America
Supplier #: A351922
Jump to:
Product Details & Documents
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
- Inner joints at bottom and side arm
- Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
Spécifications
- Bottom [ST] Joint Size:19/22
- Top ST Joint Size:[ST] 19/22
- Side ST Joint Size:19/22