Synthware Distillation Connecting Adapters
14/20, [ST] 14/20, 14/20
  60000-596
 :  A351420
Synthware Distillation Connecting Adapters
  60000-596
Synthware Distillation Connecting Adapters
Synthware Distillation Connecting Adapters
14/20, [ST] 14/20, 14/20
$91.77
Each
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
  • Inner joints at bottom and side arm
  • Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
  • Bottom [ST] Joint Size:
    14/20
  • Top ST Joint Size:
    [ST] 14/20
  • Side ST Joint Size:
    14/20