Synthware Reducing Adapters, Kemtech America | Avantor
Synthware Reducing Adapters, Kemtech America
[ST] 24/40, 45/50
Catalog # 60000-048
Supplier:   Kemtech America
Fournisseur #:  A202445
Synthware Reducing Adapters, Kemtech America
Catalog # 60000-048
Supplier:  Kemtech America
Synthware Reducing Adapters, Kemtech America
Synthware Reducing Adapters, Kemtech America
[ST] 24/40, 45/50
Price
118,07 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Reducing adapters are used for connecting dissimilar [ST] joints.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
Spécifications
  • Bottom [ST] Joint Size:
    45/50
  • Top ST Joint Size:
    [ST] 24/40