Synthware Reducing Adapters, Kemtech America
[ST] 24/40, 29/42
Catalog # 60000-044
Supplier: Kemtech America
Supplier #: A202429
Jump to:
Product Details & Documents
Reducing adapters are used for connecting dissimilar [ST] joints.
- Top outer [ST] joint
- Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
Specifications
- Bottom [ST] Joint Size:29/42
- Top ST Joint Size:[ST] 24/40