Synthware Reducing Adapters, Kemtech America
Synthware Reducing Adapters, Kemtech America
[ST] 24/40, 29/42
Catalog # 60000-044
Supplier:   Kemtech America
Supplier #:  A202429
Synthware Reducing Adapters, Kemtech America
Catalog # 60000-044
Supplier:  Kemtech America
Synthware Reducing Adapters, Kemtech America
Synthware Reducing Adapters, Kemtech America
[ST] 24/40, 29/42
Price
$76.03
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Reducing adapters are used for connecting dissimilar [ST] joints.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
Specifications
  • Bottom [ST] Joint Size:
    29/42
  • Top ST Joint Size:
    [ST] 24/40