Synthware Reducing Adapters, Kemtech America
[ST] 19/22, 24/40
Catalog # 60000-040
Supplier: Kemtech America
Fournisseur #: A201924
Jump to:
Product Details & Documents
Reducing adapters are used for connecting dissimilar [ST] joints.
- Top outer [ST] joint
- Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
Spécifications
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 19/22