Synthware Reducing Adapters, Kemtech America | Avantor
Synthware Reducing Adapters, Kemtech America
[ST] 19/22, 24/40
Catalog # 60000-040
Supplier:   Kemtech America
Fournisseur #:  A201924
Synthware Reducing Adapters, Kemtech America
Catalog # 60000-040
Supplier:  Kemtech America
Synthware Reducing Adapters, Kemtech America
Synthware Reducing Adapters, Kemtech America
[ST] 19/22, 24/40
Price
$66.97
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Reducing adapters are used for connecting dissimilar [ST] joints.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
Spécifications
  • Bottom [ST] Joint Size:
    24/40
  • Top ST Joint Size:
    [ST] 19/22