Synthware Reducing Adapters, Kemtech America
[ST] 14/20, 19/22
  60000-036
 :  A201419
Synthware Reducing Adapters, Kemtech America
  60000-036
Synthware Reducing Adapters, Kemtech America
Synthware Reducing Adapters, Kemtech America
[ST] 14/20, 19/22
$48.54
Each
Reducing adapters are used for connecting dissimilar [ST] joints.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
  • Bottom [ST] Joint Size:
    19/22
  • Top ST Joint Size:
    [ST] 14/20