Synthware Reducing Adapters, Kemtech America
[ST] 14/20, 19/22
60000-036
: A201419
Reducing adapters are used for connecting dissimilar [ST] joints.
- Top outer [ST] joint
- Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
- Bottom [ST] Joint Size:19/22
- Top ST Joint Size:[ST] 14/20