Staticide® Concentrate, ACL Staticide
Staticide® Concentrate, ACL Staticide
Drum, 189.3 L (50 gal.)
Catalog # 58611-407
Supplier:   ACL Staticide
Supplier #:  3000D
Staticide® Concentrate, ACL Staticide
Catalog # 58611-407
Supplier:  ACL Staticide
Staticide® Concentrate, ACL Staticide
Staticide® Concentrate, ACL Staticide
Drum, 189.3 L (50 gal.)
Price
$52,272.58
Log in for availability
Each (50US Gal)
Product Details & Documents
Staticide® Concentrate is available to save freight, handling, and storage costs while offering greater flexibility in dilution.
Staticide® Concentrate can be diluted with tap waater, Isopropyl Alcohol, Carbon Tetrachloride, or Perchloroethylene to meet various processing needs.
Staticide® Concentrate should be mixed with water or other solvents in the following concentrate solvent ratios to obtain these formulations: General Purpose Staticide® 1:99, Heavy Duty Staticide® 1:39
Certifications: When properly applied, Staticide® complies with or exceeds all electrostatic decay criteria established for antistatic performance as set forth by NFPA and MIL B-81705, when tested in accordance with Federal Test Std 101B, Method 4046.
Ordering information: Staticide® Concentrate is available in quart, gallon, and 50 gallon drum containers.
Specifications
  • Désignation:
    Drum
  • Taille:
    189.3 L (50 gal.)