Cube® Inserts, Hedwin | Avantor
Cube® Inserts, Hedwin
0.9 L (0.2 gal.), With Screw Cap, Natural
Catalog # 56616-415
Supplier:   Hedwin
Fournisseur #:  CUB6050
Cube® Inserts, Hedwin
Catalog # 56616-415
Supplier:  Hedwin
Cube® Inserts, Hedwin
Cube® Inserts, Hedwin
0.9 L (0.2 gal.), With Screw Cap, Natural
Price
$1,776.80
Log in pour la disponibilité
Case of 128
Product Details & Documents
Durable, lightweight cube-shaped inserts are for use with Cubitainer® cartons (see 24300-655 series) for specimen collection and food or chemical storage. Inserts may also be used for stand-alone product packaging.
Top face accomodates 38mm (11/2") or 63mm (21/2") opening, and features molded-in reversible well that can be raised for pouring or depressed for storage
Amber color insert protects contents from UV rays and is ideal for patient collection of urine samples over 24-hour period. Features integrated handle, graduation marks, and labeling tag.
Cube® material (linear low density polyethylene blend) conforms to FDA Regulation 177.1520.
Non-glug design makes contents flow in continuous, uninterrupted stream. Containers collapse as product dispenses. Inserts are supplied with screw-on cap; natural color insert is supplied with glue-on neck.
Ordering information: Inserts are shipped folded into a space-saving nesting configuration. Inserts may be fluorine-treated to prevent chemical permeation, or "PR" coated for oxygen-sensitive food or chemical products; contact your Avantor sales representative for details.
Documents
Spécifications
  • Capacité:
    0.9 L (0.2 gal.)
  • Color:
    Natural
  • Désignation:
    With Screw Cap
Accessories
Informations sur le produit
  • Catalog #
    Weight
    Availability
    Price
  • Carton, Semi-White, Double Wall
    Carton, Semi-White, Double Wall
    56616-402
    275 lbs.
    Log in pour la disponibilité
    Pack of 1 - $32.32