Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Bench Cans, Justrite® | Avantor

Bench Cans, Justrite®

0.9 L (0.25 gal.), 12.3 x 13.9 cm (47/8 x 51/2")
Catalog # 56609-720
Supplier:   Justrite
Fournisseur #:  10175
Bench Cans, Justrite®
Catalog # 56609-720
Supplier:  Justrite
Bench Cans, Justrite®
Bench Cans, Justrite®

0.9 L (0.25 gal.), 12.3 x 13.9 cm (47/8 x 51/2")

Price
$296.87
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

Terne plate steel bench can for convenient cleaning of small parts in solvents

Simply place part on spring-loaded, perforated dasher plate, press down and agitate, then release to return dasher to drain position. Plated steel dasher is an effective flame arrester. Hinged cover minimizes evaporation losses. Protected rim reduces possibility of hand injury. Item 56609-724 is a can with parts basket.

Certifications: FM approved.

Spécifications
  • Capacité:
    0.9 L (0.25 gal.)
  • O.D.×H:
    12.3 x 13.9 cm (47/8 x 51/2")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.