VWR® Microdispenser Replacement Kits
20 uL VWR® Microdispenser Replacement Kit
Catalog # 53506-912
Supplier: VWR International
Jump to:
Product Details & Documents
The 5, 10, and 20 µl VWR® Microdispenser Replacement Kits each include 2 plungers, 2 O-rings, 1 collet, and 1 coil spring and barrel tube.
- Available in a variety of sizes
Delivery information: The 20 and 25, 50, 100, 100/200, and 250 µl VWR® Microdispenser Replacement Kits each include 1 plunger, 2 O-rings, 1 collet, 3 fluoropolymer resin tips, and 1 coil spring and barrel tube.
Spécifications
- Désignation:20 uL VWR® Microdispenser Replacement Kit
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.