Disposable Pasteur Pipettes, Borosilicate Glass, Wheaton
14.6 cm (5¾")
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Fournisseur #: 357331
Disposable Pasteur Pipettes, Borosilicate Glass, Wheaton
Catalog # 53499-630
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
For use in inoculating culture media and as a general sampling pipet.
Also used in applying samples in chromatography. Straight tip can be bent over a flame. Constriction near end to facilitate plugging. Pipettes have an Ext.Ø of 7 ±0.5 mm at the top.
Ordering information: Case of 1000 Pasteur Pipettes. Each case includes 4 inner cartons of 250 pipettes.
Spécifications
- Overall Length:14.6 cm (5¾")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



