Digestion Tubes, Labconco®
250 ml, Volumetric Tubes
Catalog # 51001-204
Supplier: Labconco
Fournisseur #: 2303005
Jump to:
Product Details & Documents
Precise height and controlled wall thickness ensure exhaust-system compatibility and accurate, consistent fit.
Made of thick-wall borosilicate glass with heavy-duty retention lip and contoured bottom for optimum heat transfer and minimal thermal shock. Volumetric tubes feature volumetric marking for accurate dilutions, and built-in constriction for maximum refluxing and digestion speed.
Spécifications
- Volume:250 ml
- Désignation:Volumetric Tubes