Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Slide Holders, Jaece

Slide Holders, Jaece

Slide Holder, 17.8×30.5 cm (7×12"), 78
Catalog # 48429-105
Supplier:   Jaece
Supplier #:  L500-D
Slide Holders, Jaece
Catalog # 48429-105
Supplier:  Jaece
Slide Holders, Jaece
Slide Holders, Jaece

Slide Holder, 17.8×30.5 cm (7×12"), 78

Price
$137.62
Log in for availability
Each
Product Details & Documents

High-quality white plastic receptacle to hold slides during evaluation and cleaning.

  • Durable plastic with beveled edges
  • 39 or 78 slide capacity formats available
  • Chemical resistant
  • High-density polyethylene construction
  • Surface-griping rubber feet

These strong, lightweight slide holders are chemical resistant and provide an inexpensive way to dry slides and drain excess fluids from flat surfaces. Beveled edges provide easy access to channels and aid in slide placement or withdrawal. Inscriptions with Identi-Marker pens may be made for quick identification.

Specifications
  • Désignation:
    Slide Holder
  • Dimensions:
    17.8×30.5 cm (7×12")
  • No. of Places:
    78