Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Slide Holders, Jaece | Avantor

Slide Holders, Jaece

Slide Holder, 10.2×30.5 cm (4×12"), 39
Catalog # 48429-103
Supplier:   Jaece
Fournisseur #:  L500-C
Slide Holders, Jaece
Catalog # 48429-103
Supplier:  Jaece
Slide Holders, Jaece
Slide Holders, Jaece

Slide Holder, 10.2×30.5 cm (4×12"), 39

Price
$106.19
Log in pour la disponibilité
Each
Product Details & Documents

High-quality white plastic receptacle to hold slides during evaluation and cleaning.

  • Durable plastic with beveled edges
  • 39 or 78 slide capacity formats available
  • Chemical resistant
  • High-density polyethylene construction
  • Surface-griping rubber feet

These strong, lightweight slide holders are chemical resistant and provide an inexpensive way to dry slides and drain excess fluids from flat surfaces. Beveled edges provide easy access to channels and aid in slide placement or withdrawal. Inscriptions with Identi-Marker pens may be made for quick identification.

Spécifications
  • Désignation:
    Slide Holder
  • Dimensions:
    10.2×30.5 cm (4×12")
  • No. of Places:
    39
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.