Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Low Density Flat Poly Liners, Associated Bag

Yellow, 40 gal. cans (liner circumference 72"), 36×48", 6 mil.
  47732-648
 :  71-8-38Y
Low Density Flat Poly Liners, Associated Bag
  47732-648
Low Density Flat Poly Liners, Associated Bag
Low Density Flat Poly Liners, Associated Bag

Yellow, 40 gal. cans (liner circumference 72"), 36×48", 6 mil.

 
$626.46
   
Pack of 50
 

Sort and identify different types of waste.

Strong, bottom-sealed liners are free of heavy metals, making them safe for landfill or incineration.

 
  • Color:
    Yellow
  • Épaisseur:
    6 mil.
  • W×L:
    36×48"
  • Fits:
    40 gal. cans (liner circumference 72")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.